missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27494-1-AP-20UL
This item is not returnable.
View return policy
Description
CNPY2 is positive regulator of neurite outgrowth by stabilizing myosin regulatory light chain (MRLC). It prevents MIR-mediated MRLC ubiquitination and its subsequent proteasomal degradation.Specifications
| GTF3C2 | |
| Polyclonal | |
| Unconjugated | |
| GTF3C2 | |
| GTF3C2, KIAA0011, TF3C beta, TFIIIC 110 kDa subunit, TFIIIC BETA, TFIIIC110 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2976 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WUA4 | |
| GTF3C2 | |
| GTF3C2 Fusion Protein Ag26666 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction