missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C4 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60434FR
This item is not returnable.
View return policy
Description
GTF3C4 Polyclonal antibody specifically detects GTF3C4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GTF3C4 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.3.1.48, FLJ21002, general transcription factor 3C 4, general transcription factor 3C polypeptide 4, general transcription factor IIIC, polypeptide 4 (90kD), general transcription factor IIIC, polypeptide 4, 90kDa, KAT12, TF3C-delta, TFIII90, TFIIIC 90 kDa subunit, TFIIIC290, TFIIIC90MGC138450, TFIIICDELTA, Transcription factor IIIC 90 kDa subunit, Transcription factor IIIC subunit delta | |
| The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human general transcription factor 3C polypeptide 4 (Transcription factor IIIC 90 kDa subunit) using the numbering given in entry NP_036336.1 (GeneID 9329). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9329 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction