missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DjC9 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-97848AF594
This item is not returnable.
View return policy
Description
DjC9 Polyclonal antibody specifically detects DjC9 in Human samples. It is validated for Western Blot
Specifications
| DjC9 | |
| Polyclonal | |
| Western Blot | |
| DnaJ (Hsp40) homolog, subfamily C, member 9, dnaJ homolog subfamily C member 9 | |
| The immunogen recognized by this antibody maps to a region between residue 210 and 260 of human DnaJ (Hsp40) homolog, subfamily C, member 9 using the numbering given in entry NP_056005.12 (GeneID 23234). | |
| 0.1 mL | |
| Signal Transduction | |
| 23234 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction