missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF3C5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567074
This item is not returnable.
View return policy
Description
Immunogen sequence: KTSSQLVTMHD LKQGLGPSGT SGARKPASSK YKLKDSVYIF REGALPPYRQ MFYQLCDLNV EELQKIIHR Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.
Involved in RNA polymerase III-mediated transcription. Integral, tightly associated component of the DNA-binding TFIIIC2 subcomplex that directly binds tRNA and virus-associated RNA promoters. [UniProt].
Specifications
| GTF3C5 | |
| Polyclonal | |
| Unconjugated | |
| Gtf3c5 | |
| 2700084A09Rik; CDABP0017; General transcription factor 3C polypeptide 5; general transcription factor III polypeptide 5; general transcription factor IIIC subunit 5; general transcription factor IIIC, polypeptide 5; general transcription factor IIIC, polypeptide 5, 63kDa; GTF3C5; im:7156509; LOW QUALITY PROTEIN: general transcription factor 3C polypeptide 5; TF3C-epsilon; TFIIIC 63 kDa subunit; TFiiiC2-63; TFIIIC63; TFIIICepsilon; transcription factor IIIC 63 kDa subunit; Transcription factor IIIC subunit epsilon; transcription factor IIIC, 63 kD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9328 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5Q8 | |
| Gtf3c5 | |
| Recombinant Human GTF3C5. Recombinant protein control fragment (Product #RP-105487). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction