missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF3C6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563948
This item is not returnable.
View return policy
Description
Immunogen sequence: DKSLELEEEE IQMNDSSNLS CEQEKPMHLE IEDSGPLIDI PSETEGSVFM ETQMLP Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 41%.
Specifications
| GTF3C6 | |
| Polyclonal | |
| Unconjugated | |
| GTF3C6 | |
| bA397G5.3; C6orf51; CDA020; General transcription factor 3C polypeptide 6; general transcription factor IIIC subunit 6; general transcription factor IIIC, polypeptide 6, alpha 35kDa; GTF3C6; NPD020; TFIIIC 35 kDa subunit; TFIIIC35; Transcription factor IIIC 35 kDa subunit; transcription factor IIIC 35kDa; transcription factor IIIC subunit 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969F1 | |
| GTF3C6 | |
| Recombinant protein corresponding to Human GTF3C6. Recombinant protein control fragment (Product #RP-101825). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction