missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTPBP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569203
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 92%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 77%; Rat: 92%.
Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction.Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 [PubMed 17054726]).[supplied by OMIM].
Specifications
| GTPBP10 | |
| Polyclonal | |
| Unconjugated | |
| GTPBP10 | |
| 4930545J22Rik; BC034507; claudin 12; Cldn12; DKFZP686A10121; FLJ38242; GTP binding protein 10; GTP binding protein 10 (putative); GTP-binding protein 10; GTP-binding protein 10 (putative); GTPBP10; MGC104191; ObgH2; Protein obg homolog 2; UG0751c10; Unknown (protein for MGC:128459); zgc:92334 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 312054 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| GTPBP10 | |
| synthetic peptide directed towards the N-terminal region of rat GTPBP10. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction