missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1qA Antibody (C1QA/2955), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08748APC
This item is not returnable.
View return policy
Description
Complement Component C1qA Monoclonal antibody specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Complement Component C1qA | |
| Monoclonal | |
| APC | |
| C1QA complement component 1, q subcomponent, A chain | |
| Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) (Uniprot: P02745) | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 712 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| C1QA/2955 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction