missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atlantic Blue™ Anti-Mouse CD25/IL-2RA (PC61.5), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech AB-65137-100UG
This item is not returnable.
View return policy
Description
The CD25/IL-2RA antibody from Proteintech is a rat monoclonal antibody generated with n/a of mouse CD25/IL-2RA. This antibody recognizes mouse antigen. The CD25/IL-2RA antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.Specifications
| CD25/IL-2RA | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG1 λ |
| Flow Cytometry | |
| PC61.5 | |
| Atlantic Blue | |
| Affinity purification | |
| RUO | |
| 16184 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction