missing translation for 'onlineSavingsMsg'
Learn More

GTSE1 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen GTSE1
Clone 3B4A12
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236479
View Documents
Proteintech
68313-1-IG-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237398
View Documents
Proteintech
68313-1-IG-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The GTSE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GTSE1. This antibody recognizes human, rat, mouse antigen. The GTSE1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

GTSE1
1000 μg/mL
Monoclonal
Liquid
RUO
51512
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
3B4A12
Western Blot, ELISA
Unconjugated
Mouse
Human, Rat, Mouse
B99
IgG1
Protein G purification
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.