missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-monophosphate, Free acid, ≥98%, MP Biomedicals™

Product Code. 11304369 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 11304369

Brand: MP Biomedicals 0210190890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85-32-5
Molecular Formula C10H14N5O8P
Molecular Weight (g/mol) 363.223
InChI Key RQFCJASXJCIDSX-UUOKFMHZSA-N
Synonym 5'-guanylic acid, guanosine monophosphate, guanylic acid, guanosine-5'-monophosphate, guanosine 5'-monophosphate, 5'-gmp, guanosine 5'-phosphate, guanylate, guanidine monophosphate, guanosine-phosphate
PubChem CID 6804
ChEBI CHEBI:17345
IUPAC Name [(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)NC(=NC2=O)N

Specifications

Color White
Assay Percent Range ≥98%
Quantity 500 mg
Formula Weight 363.2
Physical Form Powder
Chemical Name or Material Guanosine-5'-monophosphate, FTIR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.