missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylate kinase Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97222CL1
This item is not returnable.
View return policy
Description
Guanylate kinase Polyclonal antibody specifically detects Guanylate kinase in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| Guanylate kinase | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1 | |
| Produced in rabbits immunized with E. coli-derived Human Guanylate kinase fragment. | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 2987 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction