missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylate kinase Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97222FR
This item is not returnable.
View return policy
Description
Guanylate kinase Polyclonal specifically detects Guanylate kinase in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Guanylate kinase | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1 | |
| Produced in rabbits immunized with E. coli-derived Human Guanylate kinase fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 2987 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction