missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylyl Cyclase alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16770
This item is not returnable.
View return policy
Description
Guanylyl Cyclase alpha 1 Polyclonal specifically detects Guanylyl Cyclase alpha 1 in Human samples. It is validated for Western Blot.
Specifications
| Guanylyl Cyclase alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EC 4.6.1.2, GC-S-alpha-1, GCS-alpha-1, GCS-alpha-3, guanylate cyclase 1, soluble, alpha 3, guanylate cyclase soluble subunit alpha-3, GUC1A3GUCA3, GUCSA3GC-SA3, GUCY1A1, Soluble guanylate cyclase large subunit | |
| Rabbit | |
| 77 kDa | |
| 0.1 mL | |
| Signal Transduction, Vision | |
| 2982 | |
| Human | |
| IgG |
| Western Blot | |
| 0.47mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q02108 | |
| GUCY1A1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human GUCY1A3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (97%), Dog (98%), Pig (97%), Rat (97%), Bovine (96%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction