Learn More
Grace Davison™ Guard Cartridge Visionht™
Brand: Grace Davison™ 5141954
Additional Details : Weight : 0.30000kg
Description
- Maximise speed and resolution
- High throughput technologies for all systems
- Simplify separation strategies
- Complementary phase chemistries
- Improve laboratory efficiency
- Seamless method transfer between UHPLC, HPLC and Prep
- UHPLC columns rated to 16,000psi for compatibility on all systems
- Low dead volume column hardware used for all UHPLC columns
- UltraMD UHPLC method development kits available containing C18 classic, C18 polar, C18 high load and C18 basic columns in selected dimension
- Other sizes available