Learn More
Invitrogen™ GUCA2A Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5121241
Product Code. 17792634
  • € 514.00 / 100µL
Description
Positive test controls include: U-251MG, Mouse small intestine, Rat small intestine. The target is usually found in the following locations: Secreted. Immunogen sequence: VTVQDGNFSF SLESVKKLKD LQEPQEPRVG KLRNFAPIPG EPVVPILCSN PNFPEELKPL CKEPNAQEIL QRLEEIAEDP GTCEICAYAA CTGC.
Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GUCA2A
Polyclonal
Unconjugated
GUCA2A
GCAP-I; GUANYL; guanylate cyclase activator 2 (guanylin 2, intestinal, heatstable); guanylate cyclase activator 2 (guanylin); guanylate cyclase activator 2A; guanylate cyclase activator 2A (guanylin 2, intestinal, heat-stable); guanylate cyclase activator 2a (guanylin); guanylate cyclase-activating protein 1; Guanylate cyclase-activating protein I; guanylin; GUCA2; Guca2a; HMW-guanylin; prepro-guanylin; STARA
Rabbit
Affinity Chromatography
RUO
14915, 25656, 2980
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.98 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P28902, P33680, Q02747
GUCA2A
Recombinant fusion protein containing a sequence corresponding to amino acids 22-115 of human GUCA2A (NP_291031.2).
100 μL
Primary
Human, Mouse, Rat