missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GUCY2C Polyclonal Antibody
GREENER_CHOICE

Product Code. 17160294 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17160294 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17160294 Supplier Invitrogen™ Supplier No. PA5112435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GUCY2C Polyclonal Antibody for Western Blot, ELISA

Receptor for the E.coli heat-stable enterotoxin (E.coli enterotoxin markedly stimulates the accumulation of cGMP in mammalian cells expressing GC-C). Also activated by the endogenous peptide guanylin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCY2C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GUCY2C
Gene Accession No. P25092, Q3UWA6
Gene Alias AI893437; DIAR6; Gcc; GC-C; guanylate cyclase 2C; guanylyl cyclase C; GUC2C; GUCY2C; heat stable enterotoxin receptor; heat-stable enterotoxin receptor; hSTAR; intestinal guanylate cyclase; MECIL; MUCIL; STA receptor; STAR
Gene Symbols GUCY2C
Host Species Rabbit
Immunogen Recombinant Human Heat-stable enterotoxin receptor protein (636-891AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14917, 2984
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.