missing translation for 'onlineSavingsMsg'
Learn More

GUCY2D Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30488490 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30488490 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30488490 Supplier Novus Biologicals Supplier No. NBP335345FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GUCY2D Polyclonal antibody specifically detects GUCY2D in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCY2D
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CORD6cone rod dystrophy 5/6, CYGD, EC 4.6.1, guanylate cyclase 2D, membrane (retina-specific), Guanylate cyclase 2D, retinal, GUC1A4, GUC2DCORD5, LCA, LCA1RCD2, retGC, RETGC1, RETGC-1EC 4.6.1.2, retinal guanylyl cyclase 1, Rod outer segment membrane guanylate cyclase, ROSGC, ROS-GC, ROS-GC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 52-350 of human GUCY2D (NP_000171.1).,, Sequence:, AVFTVGVLGPWACDPIFSRARPDLAARLAAARLNRDPGLAGGPRFEVALLPEPCRTPGSLGAVSSALARVSGLVGPVNPAACRPAELLAEEAGIALVPWGCPWTQAEGTTAPAVTPAADALYALLRAFGWARVALVTAPQDLWVEAGRSLSTALRARGLPVASVTSMEPLDLSGAREALRKVRDGPRVTAVIMVMHSVLLGGEEQRYLLEAAEELGLTDGSLVFLPFDTIHYALSPGPEALAALANSSQLRRAHDAVLTLTRHCPSEGSVLDSLRRAQERRELPSDLNLQQVSPLFGTI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3000
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.