missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GUSB Polyclonal Antibody

GUSB Polyclonal Antibody
Brand: Thermo Scientific PA5106619
This item is not returnable.
View return policy
Description
GUSB Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| GUSB | |
| Polyclonal | |
| Unconjugated | |
| GUSB | |
| Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; Gusb; Gus-r; Gus-s; Gus-t; Gus-u; Gut; lysosomal enzyme; MPS7; si:ch211-160e1.7; si:ct573103.7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110006, 24434, 2990 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P06760, P08236, P12265 | |
| GUSB | |
| A synthesized peptide derived from human GUSB(Accession P08236), corresponding to amino acid residues I326-L376. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction