missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GUSB Polyclonal Antibody
GREENER_CHOICE

Product Code. 17710554
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17710554 200 μL 200µL
17783604 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17710554 Supplier Invitrogen™ Supplier No. PA5119139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GUSB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The GUSB gene encodes beta-glucuronidase (EC 3.2.1.31), a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans. It is a tetrameric glycoprotein composed of identical subunits. The GUSB gene is mutated in mucopolysaccharidosis type VII (MPS7; MIM 253220).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUSB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GUSB
Gene Accession No. P08236
Gene Alias Ac2-223; adipose storage deficiency; AI747421; asd; beta glucuronidase; beta-D-glucuronidase; beta-G1; Beta-glucuronidase; BG; g; glucuronidase beta; glucuronidase, beta; Gur; Gus; Gusb; Gus-r; Gus-s; Gus-t; Gus-u; Gut; lysosomal enzyme; MPS7; si:ch211-160e1.7; si:ct573103.7
Gene Symbols GUSB
Host Species Rabbit
Immunogen Recombinant protein Glucuronidase Beta. The antigen corresponds to amino acid range 451-649 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2990
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.