missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GW 9662
GREENER_CHOICE

Product Code. 18034172
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18034172

Brand: Tocris Bioscience™ 1508/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective covalent PPARγ antagonist

GW 9662 is a selective irreversible PPARγ antagonist (IC50 values are 3.3, 32 and 2000 nM for PPARγ, PPARα and PPARδ respectively). Blocks the inhibition of osteoclast formation induced by IL-4 in the low micromolar range (1-2 μM), therefore is more potent than BADGE. Anticancer, inhibits growth of human mammary tumor cell lines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22978-25-2
Molecular Formula C13H9ClN2O3

Specifications

Quantity 50 mg
Formula Weight 276.68
Purity >98%
Molecular Weight (g/mol) 276.676
Chemical Name or Material GW 9662
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.