missing translation for 'onlineSavingsMsg'
Learn More

GW182 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-28751AF350

Product Code. 18488194

  • € 681.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GW182 Polyclonal antibody specifically detects GW182 in Human, Mouse samples. It is validated for Western Blot, Immunoblot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GW182
Polyclonal
Western Blot, Immunoblotting
CAG repeat protein 26, CAGH26FLJ22043, DKFZp666E117, EMSY interactor protein, Glycine-tryptophan protein of 182 kDa, GW182 autoantigen, GW182GW1, KIAA1460EDIE, MGC75384, Protein GW1, TNRC6, trinucleotide repeat containing 6, trinucleotide repeat containing 6A, trinucleotide repeat-containing gene 6A protein
The immunogen recognized by this antibody maps to a region between residue 575 and 625 of human GW182/glycine-tryptophan protein of 182 kDa (NP_055309.2).
0.1 mL
Primary
Human, Mouse
Purified
Western Blot, Immunoblot
Alexa Fluor 350
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
27327
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.