missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GW182 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28751
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GW182 Polyclonal specifically detects GW182 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunoblotting, Knockdown Validated.
Tekniske data
| GW182 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| CAG repeat protein 26, CAGH26FLJ22043, DKFZp666E117, EMSY interactor protein, Glycine-tryptophan protein of 182 kDa, GW182 autoantigen, GW182GW1, KIAA1460EDIE, MGC75384, Protein GW1, TNRC6, trinucleotide repeat containing 6, trinucleotide repeat containing 6A, trinucleotide repeat-containing gene 6A protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 27327 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation, Immunoblot | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 6 ug/mg lysate, Immunoblotting | |
| Q8NDV7 | |
| TNRC6A | |
| The immunogen recognized by this antibody maps to a region between residue 575 and 625 of human GW182/glycine-tryptophan protein of 182 kDa (NP_055309.2). | |
| 100 μL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee, White-tufted-ear marmoset and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion