missing translation for 'onlineSavingsMsg'
Learn More

GW2580, MedChemExpress

Product Code. 16454288
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16454288

Brand: Medchem Express HY10917

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW2580 is an orally bioavailable and selective inhibitor of c-Fms kinase which completely inhibits human cFMS kinase in vitro at 0.06 μM. GW2580 acts as a competitive inhibitor of ATP binding to the cFMS kinase and inhibits colony-stimulating-factor-1 signaling.

GW2580 is an orally bioavailable and selective inhibitor of c-Fms kinase which completely inhibits human cFMS kinase in vitro at 0.06 μM. GW2580 acts as a competitive inhibitor of ATP binding to the cFMS kinase and inhibits colony-stimulating-factor-1 signaling.

Specifications

Chemical Name or Material GW2580
CAS 870483-87-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22N4O3
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (90.96 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
SMILES NC1=NC(N)=C(CC2=CC(OC)=C(OCC3=CC=C(OC)C=C3)C=C2)C=N1
Molecular Weight (g/mol) 366.41
Formula Weight 366.41
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.