Learn More
A&D Instruments GX Series - Internal Calibration 210g x 0.001g Class II Approved
GREENER_CHOICE
Brand:  A&D Instruments GX-200-EC
GX Series - Internal Calibration 210g x 0.001g Class II Approved
Product Code. 16691142
  • ONLINE EXCLUSIVE PRICE
    € 1947.75
    € 1427.95 / Each
    Save € 519.80
    26% Off