missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GYPA, Mouse anti-Human, CF-Blue, Clone: HI264, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15411.100Reactions
This item is not returnable.
View return policy
Specifications
| GYPA | |
| Monoclonal | |
| CF-Blue | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CD235a/GPA/GPErik/GPSAT/GpMiIII/HGpMiIII/HGpMiV/HGpMiX/HGpMiXI/HGpSta(C)/MN/MNS | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| HI264 | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| glycophorin A (MNS blood group) | |
| GYPA | |
| A synthetic peptide corresponding to N-terminus of human GYPA. | |
| RUO | |
| 2993 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction