missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GYPE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
€ 164.00 - € 413.00
Specifications
Antigen | GYPE |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17702884
|
Invitrogen
PA5121482 |
20 μL |
€ 164.00
20µL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17792874
|
Invitrogen
PA5121481 |
100 μL |
€ 413.00
100µL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
GYPE (Glycophorin E (MNS Blood Group)) is a Protein Coding gene. Diseases associated with GYPE include Malaria and Hemidystonia. An important paralog of this gene is GYPA. The protein encoded by this gene is a sialoglycoprotein and a type I membrane protein. It is a member of a gene family with GPA and GPB genes. This encoded protein might carry the M blood group antigen. GYPA, GYPB, and GYPE are organized in tandem on chromosome 4. This gene might have derived from an ancestral gene common to the GPB gene by gene duplication. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.Specifications
GYPE | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P15421 | |
2996 | |
Ser20-Ile70 (Accession P15421), with N-terminal His and GST Tag | |
Primary | |
-20°C | |
GYPE |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Glycophorin E | |
GYPE | |
IgG | |
Antigen affinity chromatography | |
Antibody |