missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GYS2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17228063 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17228063 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17228063 Supplier Invitrogen™ Supplier No. PA5115297

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Glycogen synthase 2.

The protein encoded by this gene, liver glycogen synthase, catalyzes the rate-limiting step in the synthesis of glycogen - the transfer of a glucose molecule from UDP-glucose to a terminal branch of the glycogen molecule. Mutations in this gene cause glycogen storage disease type 0 (GSD-0) - a rare type of early childhood fasting hypoglycemia with decreased liver glycogen content.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GYS2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GYS2
Gene Accession No. P54840
Gene Alias BC021322; cb765; Glycogen [starch] synthase, liver; glycogen liver; glycogen synthase; glycogen synthase 2; glycogen synthase 2 (liver); glycogen synthase, liver; GLYSN; GYS2; LGS; zgc:112057
Gene Symbols GYS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GYS2(Accession P54840), corresponding to amino acid residues L601-A651.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2998
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.