missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H1F0 Antibody (AE-4), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08697PE
This item is not returnable.
View return policy
Description
H1F0 Monoclonal antibody specifically detects H1F0 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| H1F0 | |
| Monoclonal | |
| PE | |
| H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0 | |
| Nuclei of human leukemia biopsy cells | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| AE-4 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3005 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction