missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H1F0 Antibody (rHH1/8702), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20985H
This item is not returnable.
View return policy
Description
H1F0 Monoclonal antibody specifically detects H1F0 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| H1F0 | |
| Monoclonal | |
| HRP | |
| H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0 | |
| Nuclei of human leukemia biopsy cells | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Epigenetics, Nuclear Envelope Markers | |
| 3005 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rHH1/8702 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction