missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ H1FX Polyclonal Antibody
GREENER_CHOICE

Product Code. 16669794
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16669794 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16669794 Supplier Invitrogen™ Supplier No. PA5103345

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Histone H1X.

Histones H1 are necessary for the condensation of nucleosome chains into higher-order structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H1FX
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene H1-10
Gene Accession No. Q92522
Gene Alias H1 histone family member X; H1 histone family, member X; H1.10; H1FX; H1X; Histone H1.X; histone H1x
Gene Symbols H1-10
Host Species Rabbit
Immunogen A synthesized peptide derived from human H1FX(Accession Q92522), corresponding to amino acid residues T13-N63.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8971
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.