missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
H1N1 Neuraminidase Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41110AF700
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
H1N1 Neuraminidase Polyclonal antibody specifically detects H1N1 Neuraminidase in Influenza Virus N1, Virus samples. It is validated for ELISA
Specifikationer
| H1N1 Neuraminidase | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Antibody was raised against synthetic peptides from the seasonal influenza (H1N1) Neuraminidase protein and the novel swine influenza Neuraminidase protein. The antibody recognizes both variants. The immunogen is located within the first 50 amino acids of H1N1 Neuraminidase. Amino Acid Squence: SIQLGNQNQIET | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Influenza virus N1, Virus | |
| Purified |
| ELISA | |
| Alexa Fluor 700 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering