missing translation for 'onlineSavingsMsg'
Learn More

H1N1 Neuraminidase Antibody - BSA Free, Novus Biologicals™

Product Code. p-200074549 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18620636 0.025 mg 0.03mg
18199938 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18620636 Supplier Novus Biologicals Supplier No. NBP2411100.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

H1N1 Neuraminidase Polyclonal specifically detects H1N1 Neuraminidase in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H1N1 Neuraminidase
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Host Species Rabbit
Immunogen Antibody was raised against synthetic peptides from the seasonal influenza (H1N1) Neuraminidase protein and the novel swine influenza Neuraminidase protein. The antibody recognizes both variants. The immunogen is located within the first 50 amino acids of H1N1 Neuraminidase. Amino Acid Squence: SIQLGNQNQIET
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody will recognize the Neuraminidase from the both the seasonal influenza (H1N1) and swine-origin influenza virus (A/California/14/2009 (H1N1)).
Target Species Influenza virus, Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.