missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35820-20ul
This item is not returnable.
View return policy
Description
H2A Polyclonal antibody specifically detects H2A in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| H2A | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| H2A histone family, member O, H2A.2, H2A/O, H2A/q, H2a-615, H2AFOHistone H2A/o, HIST2H2AAH2A, histone 2, H2aa, histone 2, H2aa3, histone cluster 2, H2aa3, histone H2A type 2-A, Histone H2A.2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-208 of mouse H2A (NP_034508.2).,, Sequence:, IEADHVGTYGISVYQSPGDIGQYTFEFDGDELFYVDLDKKETVWMLPEFGQLASFDPQGGLQNIAVVKHNLGVLTKRSNSTPATNEAPQATVFPKSPVLLGQPNTLICFVDNIFPPVINITWLRNSKSVADGVYETSFFVNRDYSFHKLSYLTFIPSDDDIYDCKVEHWGLEEPVLKHWEPE | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8337 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction