missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2AFX, Mouse anti-Human, Clone: 8H4-1H11-F8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15282.100uL
This item is not returnable.
View return policy
Specifications
| H2AFX | |
| Monoclonal | |
| Unconjugated | |
| In PBS (50% glycerol, 0.5% BSA, 0.02% sodium azide). | |
| H2A.X/H2A/X/H2AX | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Western Blot | |
| 8H4-1H11-F8 | |
| Immunofluorescence (1:400) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| H2A histone family, member X | |
| H2AFX | |
| Synthetic phosphopeptide corresponding to residues surrounding S139 of human histone H2AX. | |
| RUO | |
| 3014 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction