missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2AFX, Rabbit anti-Human, FITC, Clone: 1E4, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB18902.10Reactions
This item is not returnable.
View return policy
Specifications
| H2AFX | |
| Monoclonal | |
| FITC | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| H2A.X/H2A/X/H2AX | |
| Rabbit | |
| 10 Reactions | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 1E4 | |
| Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| H2A histone family, member X | |
| H2AFX | |
| A synthetic phosphopeptide corresponding to residues surrounding S139 of human H2AFX. | |
| RUO | |
| 3014 | |
| Store at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction