missing translation for 'onlineSavingsMsg'
Learn More

H2AFY2 Monoclonal Antibody (OTI1F6), TrueMAB™, OriGene

Product Code. p-200017072
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17582543 30 μL 30µL
17562543 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17582543 Supplier Origene Technologies Supplier No. TA803459S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

H2AFY2 Monoclonal antibody specifically detects H2AFY2 in Human samples. It is validated for Western Blot

Specifications

Antigen H2AFY2
Applications Western Blot
Classification Monoclonal
Clone OTI1F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ITGB3BP
Gene Accession No. Q13352
Gene Alias Core histone macro-H2A.2; core histone macroH2A2.2; H2A histone family member Y2; H2A histone family, member Y2; H2AFY2; H2afy3; Histone macroH2A2; histone mH2A2; macroH2A2; mH2A2; RGD1561371
Host Species Mouse
Immunogen Full length human recombinant protein of human ITGB3BP produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23421
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.