missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H2BK5ac Recombinant Rabbit Monoclonal Antibody (RM455)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544553
This item is not returnable.
View return policy
Description
This antibody reacts to the Histone H3.3 G34V mutant. No cross reactivity with wild type Histone H3.3.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
Specifications
| H2BK5ac | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| A0A2R8Y619, O60814, P06899, P0C1H6, P23527, P33778, P57053, P58876, P62807, Q16778, Q5QNW6, Q7Z2G1, Q8N257, Q93079, Q96A08, Q99877, Q99879, Q99880 | |
| H2B, H2bc1, H2BC10, H2BC11, H2BC12, H2BC13, H2BC14, H2BC15, H2BC17, H2bc18, H2BC21, H2BC3, H2BC4, H2BC5, H2BC8, H2BC9, H2BE1, H2BU1, H2BW1, H2BW2, LOC102724334 | |
| An acetyl-peptide corresponding to Acetyl-Histone H2B (Lys5). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Luminex | |
| RM455 | |
| Unconjugated | |
| H2BW2 | |
| 2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K5; Acetyl-Histone H2B Lys5; AV127319; bA317E16.3; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; GL105; H2B; H2B 291A; H2B 291B; H2b 613; H2B GL105; H2B histone family member M; H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member S; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B K; H2B type 12; H2B.1; H2B.1A; H2B.1B; H2B.2; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h; H2B/I; H2B/j; H2B/k; H2B/l; H2B/n; H2B/q; H2B/r; H2B/S; H2b-143; h2B-221; H2b-291b; H2b-613; H2Bb; H2BC10; H2bc4; H2BC6; H2BC7; H2BC8; H2b-f; H2BFA; H2BFAiii; H2BFB; H2BFC; H2BFD; H2BFE; H2BFF; H2BFG; H2BFH; H2BFJ; H2BFK; H2BFL; H2BFM; H2BFN; H2BFQ; H2BFR; H2bfs; H2BFT; H2BFU; H2BFWT; H2BGL105; H2BJ; H2b-j; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2b-l; H2BLys5ac; H2b-n; H2BQ; H2BS14; H2BS14p; HIRA-interacting protein 1; HIRA-interacting protein 2; HIRIP1; HIRIP2; Hist1h2ba; Hist1h2bb; HIST1H2BC; HIST1H2BD; Hist1h2be; Hist1h2bf; Hist1h2bg; HIST1H2BH; HIST1H2BI; HIST1H2BJ; HIST1H2BK; Hist1h2bl; Hist1h2bm; Hist1h2bn; HIST1H2BO; HIST1H3I; Hist2h2be; HIST2H2BF; Hist3h2bb; histone 1, H2ba; histone 1, H2bb; histone 1, H2bc; histone 1, H2bd; histone 1, H2bg; histone 1, H2bh; histone 1, H2bi; histone 1, H2bj; histone 1, H2bk; histone 1, H2bl; histone 1, H2bm; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; Histone 2B; histone 3, H2bb; histone cluster 1, H2ba; histone cluster 1, H2bb; histone cluster 1, H2bc; histone cluster 1, H2bd; histone cluster 1, H2bg; histone cluster 1, H2bh; histone cluster 1, H2bi; histone cluster 1, H2bj; histone cluster 1, H2bk; histone cluster 1, H2bl; histone cluster 1, H2bm; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2be; histone cluster 2, H2bf; histone cluster 3, H2bb; histone family member; histone H2B; histone H2B type 1-A; Histone H2B type 1-B; histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; histone H2B type 1-D; Histone H2B type 1-F/J/L; histone H2B type 1-H; Histone H2B type 1-J; histone H2B type 1-K; Histone H2B type 1-L; histone H2B type 1-M; Histone H2B type 1-N; histone H2B type 1-O; Histone H2B type 2-E; histone H2B type 2-F; Histone H2B type 3-B; histone H2B type F-M; Histone H2B type F-S; histone H2B type F-S-like; histone H2B type W-T; Histone H2B, testis; Histone H2B.1; histone H2B.1 A; Histone H2B.1 B; histone H2B.2; Histone H2B.a; histone H2B.b; Histone H2B.c; histone H2B.d; Histone H2B.e; histone H2B.f; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.j; histone H2B.k; Histone H2B.l; histone H2B.n; Histone H2B.q; histone H2B.r; Histone H2B.s; histone H2B-GL105; LOC102724334; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; STBP; T25626; testis-specific histone 2b; testis-specific histone H2B; Th2b; TSH2B; TSH2B.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 102724334, 114483833, 128312, 158983, 255626, 286436, 3017, 3018, 440689, 8339, 8340, 8341, 8342, 8345, 8346, 8347, 8348, 8349, 85236, 8970 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction