missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H3.3 G34W oncohistone mutant Recombinant Rabbit Monoclonal Antibody (RM263), Biotin

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544563
This item is not returnable.
View return policy
Description
This antibody reacts to native human PSA (Prostate Specific Antigen), including both free (unbound) form and in a complex with ACT (alpha 1-antichymotrypsin). It may react to reduced PSA at a higher concentration.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3.
Specifications
| H3.3 G34W oncohistone mutant | |
| Recombinant Monoclonal | |
| 250 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| P84243 | |
| H3-3A, H3-3B | |
| A peptide corresponding to Histone H3.3 G34W mutant. The immunogen range is a synthetic peptide within amino acids 1-100 of histone H3.3 with G34W mutant. | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| RM263 | |
| Biotin | |
| H3-3A | |
| H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3; H3.3 G34W; H3.3A; H3.3B; H3/g; H3-3A; H3-3B; H3B; H3F3; H3F3A; H3F3B; H3FT; H3G34W; H3t; Histone 3.3; Histone cluster 3; Histone H3; Histone H3.3; Histone H3.3 G34W; Histone H3.3G34W; PP781 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3020, 3021 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction