missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H4 Clustered Histone 1 [ac Lys8] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 724.00
Specifications
| Antigen | H4 Clustered Histone 1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation -seq 1:20 - 1:50, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:200 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ChIP Assay, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230380
|
Novus Biologicals
NBP3-38403-100ul |
100 μL |
€ 724.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227786
|
Novus Biologicals
NBP3-38403-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
H4 Clustered Histone 1 Polyclonal antibody specifically detects H4 Clustered Histone 1 in Human,Mouse,Rat samples. It is validated for ChIP assay,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifica
| H4 Clustered Histone 1 | |
| ChIP Assay, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| H4-16;H4C11;H4C12;H4C13;H4C14;H4C15;H4C2;H4C3;H4C4;H4C5;H4C6;H4C8;H4C9;H4FA, H4F4, HIST1H4A | |
| A synthetic acetylated peptide around K8 of human H4 Clustered Histone 1 (NP_003529.1).,, Sequence:, GGKGL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation -seq 1:20 - 1:50, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:200 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 8359 | |
| IgG | |
| Affinity purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto