missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H4K20ac Recombinant Rabbit Monoclonal Antibody (RM205), ChIP-Verified
Rabbit Recombinant Monoclonal Antibody
€ 143.00 - € 552.00
Specifications
| Antigen | H4K20ac |
|---|---|
| Clone | RM205 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| Applications | ChIP Assay, ELISA, Western Blot, Luminex, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17119295
|
Invitrogen™
MA533387 |
100 μg |
€ 552.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17135283
|
Invitrogen™
MA53338720UG |
20 μg |
€ 143.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Click here for Master Lot linking information.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.Specifications
| H4K20ac | |
| 1 mg/mL | |
| ChIP Assay, ELISA, Western Blot, Luminex, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| P62805 | |
| 121504, 8294, 8359, 8363, 8366, 8367, 8370 | |
| An acetyl-peptide corresponding to Acetyl-Histone H4 (Lys20). | |
| Antibody |
| RM205 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Yeast | |
| H4C1 | |
| acetyl Histone H4; Acetyl-H4 K20; Acetyl-H4 Lys20; Acetyl-Histone H4 K20; Acetyl-Histone H4 Lys20; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K20ac; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/j; H4/K; H4/M; H4/n; H4/O; H4/p; H4-16; H4C1; H4C11; H4C12; H4C13; H4C14; H4C15; H4C2; H4C3; H4C4; H4C5; H4C6; H4C8; H4C9; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K20; H4K20a; H4Lys20ac; H4M; HIST1H4; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; HIST2H4; HIST2H4A; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; histone H4; Histone H4 acetyl; Histone H4A; Histone H4K20; Histone H4K20ac; histone IV, family 2 | |
| H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9 | |
| Primary | |
| Protein A |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts