missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H4K5me1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144313
This item is not returnable.
View return policy
Description
H4K5me1 Polyclonal Antibody for ICC/IF, ChIP, ELISA
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
| H4K5me1 | |
| Polyclonal | |
| Unconjugated | |
| HIST1H4A | |
| H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4-16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; HIST2H4; HIST2H4A; HIST2H4B; HIST4H4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 121504 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, ELISA, Immunocytochemistry | |
| 0.08 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P62805 | |
| HIST1H4A | |
| Peptide sequence around site of Mono-methyl-Lys (5) derived from Human Histone H4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction