missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H4R3me1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116986
This item is not returnable.
View return policy
Description
Positive Samples: HeLa Immunogen sequence: SGRGK.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
| H4R3me1 | |
| Polyclonal | |
| Unconjugated | |
| H4C1 | |
| FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 R3; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/j; H4/K; H4/M; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4Arg3me; H4Arg3me1; H4C1; H4c11; H4c12; H4C13; H4C14; H4C15; H4c2; H4c3; H4c4; H4C5; H4c6; H4c8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4M; H4R3; H4R3me; HIST1H4; HIST1H4A; Hist1h4b; HIST1H4C; Hist1h4d; HIST1H4E; HIST1H4F; HIST1H4H; Hist1h4i; HIST1H4J; Hist1h4k; HIST1H4L; Hist1h4m; Hist2h4; Hist2h4a; HIST2H4B; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; histone H4; Histone H4A; Histone H4R3me1; histone IV, family 2; methyl Histone 3; methyl Histone H3; Methyl-H4 Arg3; Methyl-H4 R3; Methyl-Histone H4 Arg3; Methyl-Histone H4 R3; Mono-methyl H4; Mono-methyl Histone H4; X04652 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 121504, 319158, 326619, 326620, 679983, 8294, 8359, 8363, 8366, 8367, 8370, 97122 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4.06 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P62804, P62805, P62806 | |
| H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9, HIST1H4A | |
| A synthetic monomethylated peptide around R3 of human Histone H4 (NP_0035291). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction