missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H60 Antibody (RM0248-11G35), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12428PE
This item is not returnable.
View return policy
Description
H60 Monoclonal specifically detects H60 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| H60 | |
| Monoclonal | |
| PE | |
| H60a | |
| Mouse recombinant protein of H60. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| RM0248-11G35 | |
| Flow Cytometry | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 15101 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction