missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H60 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547081
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Enables natural killer cell lectin-like receptor binding activity. Acts upstream of or within several processes, including positive regulation of immune response; positive regulation of interferon-gamma production; and positive regulation of leukocyte activation.
Specifications
| H60 | |
| Polyclonal | |
| Unconjugated | |
| H60a | |
| H60; H60a; histocompatibility 60; histocompatibility 60a | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 15101 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| 0 | |
| H60a | |
| Mouse myeloma cell line NS0-derived recombinant mouse H60 Asp30-Gln212. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction