missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H6PD Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552142
This item is not returnable.
View return policy
Description
Immunogen sequence: APMPSDFQVL RAKYRESPLV SAWSEELISK LANDIEATAV RAVRRFGQFH LALSGGSSPV ALFQQLATAH YGFPWAHTHL WLVDERCVPL SDPESNFQGL QAHLLQHVRI PYYNIHP Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.
Specifications
| H6PD | |
| Polyclonal | |
| Unconjugated | |
| H6PD | |
| 6PGL; 6-phosphogluconolactonase; AI785303; CORTRD1; EGK_00198; G6PD, H form; G6pd1; G6PDH; GDH; GDH/6PGL endoplasmic bifunctional protein; glucose 1- dehydrogenase; glucose 1-dehydrogenase; glucose dehydrogenase; glucose dehyrogenase; Glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase 1; glucose-6-phosphate dehydrogenase, salivary; Gpd1; Gpd-1; H6pd; H6pdh; hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase); hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100198, 9563 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95479, Q8CFX1 | |
| H6PD | |
| Recombinant protein corresponding to Human H6PD. Recombinant protein control fragment (Product #RP-102368). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction