Learn More
HA Tag Monoclonal Antibody (2-2.2.14), DyLight™ 550, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 26183D550
Additional Details : Weight : 0.00100kg
Description
26183-D550 has been successfully used for immunofluorescence. DyLight 550 has an excitation/emission of 562/576 nm. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
HA Tag Monoclonal antibody specifically detects HA Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.Specifications
HA Tag | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Immunocytochemistry, Western Blot | |
2-2.2.14 | |
DyLight 550 | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
HA peptide YPYDVPDYA derivitized to ovalbumin. | |
50 μL | |
Primary | |
Antibody | |
IgG1 |