Learn More
Thermo Scientific™ HAAKE™ Viscotester™ 3 Rotational Viscometer
Product Code. 16689592
Change view
Click to view available options
Viscosity Range:
1.5 to 1300 mPas
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Unit Size
16689592 1.5 to 1300 mPas Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16689592 Supplier Thermo Scientific™ Supplier No. 3990111
  • ONLINE EXCLUSIVE PRICE
    € 3250.00
    € 2995.00 / Pack of 1
    Save € 255.00
    7% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ingenious design makes this portable viscometer the easiest you’ll ever use for fast, reliable measurements anywhere in your process.
Perform viscosity measurements with ease. The Thermo Scientific™ HAAKE™ Viscotester™ 3 rotational viscometer ensures quick, reliable tests for QC applications.
Use it as a handheld viscometer or on a lab stand. Color coding ensures measuring geometries match the analysis, the display is readable in any lighting, and the viscometer remembers the reference point measurement for you. All that comes in a ready-to-go carrying case.
HAAKE Viscotester 3 Rotational Viscometer is the replacement for the discontinued Thermo Scientific™ HAAKE™ Viscotester™ 1 plus and 2 plus Rotational Viscometers.
Designed for simplicity – The HAAKE Viscotester 3 rotational viscometer is built for ease of use to get you quickly to the right viscosity measurements:
  • Self-explaining handling
  • Smart alignment given by design and visualized by an electronic level
  • Quick coupling of measuring geometries for fast and easy adaptation
  • Color coded geometries and a geometry range indicator ensure correct rotor selection every time
  • Adaptable display brightness ensures ideal readability in any environment
  • Battery and power supply options allow the viscometer to be run anywhere
  • Memory assist functionality allows you to quickly and easily compare measured viscosities with the reference point
  • Selectable viscosity units (mPa s or dPa s)
  • Compatibility with previous viscometer models
Recommended for: your entire process, from accepting the delivery of goods, to sampling during production and quality control (QC).
  • Printing inks, paints, inks and other coatings
  • Shampoos, creams, lotions, and similar personal care products
  • Oil, greases, pastes and similar substances
  • Sauces, thickeners, and similar food products
TRUSTED_SUSTAINABILITY
Specifications
Description HAAKE Viscotester 3 Rotational Viscometer for low viscosity measurements
Includes Base viscometer comes in carrying case with instrument holder (for lab stand), two measuring cups (A and B), three rotors (No. 3, 4 and 5), four rechargeable batteries and one power supply adapter. Easily upgrade your Thermo Scientific HAAKE Viscotester 1 Plus viscometer to this model; the results are compatible.
Measuring Accuracy ± 5% FSD (rotors R1, R2, R3)
± 7.5% FSD (rotor R5)
± 10% FSD (rotor R4)
Power Supply Operable by battery or power supply.
Rotational Speed Range 62.5 rpm
Temperature Range (Metric) Up to 150°C
Viscosity Range 1.5 to 1300 mPas