missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HABP1/C1QBP/GC1q R Antibody (1G7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42691JF549
This item is not returnable.
View return policy
Description
HABP1/C1QBP/GC1q R Monoclonal antibody specifically detects HABP1/C1QBP/GC1q R in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| HABP1/C1QBP/GC1q R | |
| Monoclonal | |
| Janelia Fluor 549 | |
| C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein | |
| Recombinant human HABP1/C1QBP/GC1q R (74-282aa) purified from E.coli. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 708 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 1G7 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction