missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HABP2 Monoclonal Antibody (OTI5H3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527128
This item is not returnable.
View return policy
Description
HABP2 Monoclonal Antibody for Western Blot, IHC (P)
HABP2 is an extracellularserine protease that binds hyaluronic acid and is involved in celladhesion. The encoded protein is synthesized as a single chain, butthen undergoes an autoproteolytic event to form the functionalheterodimer. Further autoproteolysis leads to smaller, inactivepeptides. This protease is known to cleave urinary plasminogenactivator, coagulation factor VII, and the alpha and beta chains offibrinogen, but not prothrombin, plasminogen, or the gamma chain offibrinogen. Two transcript variants encoding different isoformshave been found for this gene.
Specifications
| HABP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14520 | |
| HABP2 | |
| Human recombinant protein fragment corresponding to amino acids 33-330 of HABP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5H3 | |
| Unconjugated | |
| HABP2 | |
| AI035669; Factor seven-activating protease; factor VII activating protein; factor VII-activating protease; FSAP; HABP; Habp2; Hepatocyte growth factor activator-like protein; HGFAL; hyaduronic acid-binding protein 2; hyaluronan binding protein 2; hyaluronan-binding protein 2; Hyaluronan-binding protein 2 27 kDa light chain; Hyaluronan-binding protein 2 27 kDa light chain alternate form; Hyaluronan-binding protein 2 50 kDa heavy chain; Hyaluronan-binding protein 2 50 kDa heavy chain alternate form; hyaluronic acid binding protein 2; NMTC5; PHBP; plasma hyalurona; plasma hyaluronan binding protein; plasma hyaluronan-binding protein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3026 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction