missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HABP2 Monoclonal Antibody (OTI8D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809299
This item is not returnable.
View return policy
Description
HABP2 is an extracellularserine protease that binds hyaluronic acid and is involved in celladhesion. The encoded protein is synthesized as a single chain, butthen undergoes an autoproteolytic event to form the functionalheterodimer. Further autoproteolysis leads to smaller, inactivepeptides. This protease is known to cleave urinary plasminogenactivator, coagulation factor VII, and the alpha and beta chains offibrinogen, but not prothrombin, plasminogen, or the gamma chain offibrinogen. Two transcript variants encoding different isoformshave been found for this gene.Specifications
| HABP2 | |
| Monoclonal | |
| Unconjugated | |
| HABP2 | |
| FSAP, HABP, HGFAL, NMTC5, PHBP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3026 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8D6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14520 | |
| HABP2 | |
| Human recombit protein fragment corresponding to amino acids 33-330 of human HABP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction